Tisk…

Methyl phenylpropiolate 98%

Dodavatel: Thermo Scientific

Synonyms: Methyl Phenylpropiolate, Phenylpropiolic acid methyl ester

334590050 334590250
ACRO334590050EA 2320 CZK
ACRO334590050 ACRO334590250
Methyl phenylpropiolate 98%
Methyl phenylpropiolate
Vzorec: C₁₀H₈O₂
MW: 160,17 g/mol
Bod varu: 109…110 °C (2 mmHg)
Bod tání: 26 °C
Hustota: 1,083 g/cm³ (25 °C)
MDL Number: MFCD00041685
CAS číslo: 4891-38-7

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5615 to 1.5645 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR