Chromium hexacarbonyl 99+%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Chromium carbonyl, Hexacarbonylchromium, Hexacarbonylchromium(0), Chromium(0) hexacarbonyl
221050100
221050500
ACRO221050100EA
4970
CZK
ACRO221050100
ACRO221050500
Chromium hexacarbonyl 99+%
Chromium hexacarbonyl
Vzorec:
C₆CrO₆ MW: 220.06 Bod varu: 220 °C (760 mmHg) Bod tání: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010945 CAS číslo: 13007-92-6 EINECS: 235-852-4 UN: 3466 ADR: 6.1 III Merck Index: 13,02253 |
Výsledky specifikačního testu
Appearance | White to yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Elemental Analysis | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...