Tisk…

o-Phenylenediamine dihydrochloride 99+%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Benzene-1,2-diamine dihydrochloride, 1,2-Phenylenediammonium dichloride, 1,2-Diaminobenzene dihydrochloride, 1,2-Phenylenediamine dihydrochloride

218480250 218481000
ACRO218480250EA 3250 CZK
ACRO218480250 ACRO218481000
o-Phenylenediamine dihydrochloride 99+%
o-Phenylenediamine dihydrochloride
Vzorec: C₆H₁₀Cl₂N₂
MW: 181,06 g/mol
Bod tání: 258 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00012966
CAS číslo: 615-28-1
EINECS: 210-418-7
UN: 2811
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance Light pink to purple Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR