Dipotassium tetrachloropalladate > 32,0% Pd
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Potassium tetrachloropalladate (II), Potassium chloropalladite
195120020
195120100
ACRO195120020EA
7800
CZK
ACRO195120020
ACRO195120100
Dipotassium tetrachloropalladate > 32,0% Pd
Dipotassium tetrachloropalladate
Vzorec:
Cl₄K₂Pd MW: 326,43 g/mol Bod tání: 105 °C Hustota: 2,67 g/cm³ |
MDL Number:
MFCD00011373 CAS číslo: 10025-98-6 EINECS: 233-049-3 |
Výsledky specifikačního testu
Appearance | Green-brown to dark brown Crystalline powder or crystals |
Assay | ≥32.0 % (Pd) |
Calcium (Ca) | ≤10 ppm |
Copper (Cu) | ≤10 ppm |
Gold (Au) | ≤20 ppm |
Iridium (Ir) | ≤20 ppm |
Iron (Fe) | ≤10 ppm |
Magnesium (Mg) | ≤10 ppm |
Platinum (Pt) | ≤20 ppm |
Rhodium (Rh) | ≤20 ppm |
Ruthenium (Ru) | ≤20 ppm |
Silicon (Si) | ≤30 ppm |
Silver (Ag) | ≤20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...