Tisk…

Cottonseed oil, pure

Dodavatel: Thermo Scientific

177115000 177110025
ACRO177115000EA 1730 CZK
ACRO177115000 ACRO177110025
Cottonseed oil, pure
Cottonseed oil
Hustota: 0,91 g/cm³
Bod vzplanutí: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00130872
CAS číslo: 8001-29-4
EINECS: 232-280-7

Objednat

Výsledky specifikačního testu

Appearance Yellow Oily liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4730 (20°C, 589 nm)
Additional info linolenic and higher: ≤1.5%
Acid value ≤0.5 mg KOH/g
Iodine value 95 to 115 g I2/100 g
Saponification value 185 to 198 mg KOH/g
Unsaponifiable matter ≤1.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR