Phthalimide potassium salt derivative 99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
N-Potassium phthalimide, Potassium 1,3-dioxoisoindolin-2-ide, Potassium phthalimide
170861000
170865000
170860025
ACRO170861000EA
1700
CZK
ACRO170861000
ACRO170865000
ACRO170860025
Phthalimide potassium salt derivative 99%
Phthalimide potassium salt
Vzorec:
C₈H₄KNO₂ Bod tání: >300 °C Hustota: 1,634 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005887 CAS číslo: 1074-82-4 EINECS: 214-046-6 |
Výsledky specifikačního testu
Appearance | White to yellow to green Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % (K.F.) |
Titration with HClO4 | ≥98.5 % (On dried substance) |
HPLC | ≥97.5 % |
Specific optical rotation | -63° to -67° (20°C, 589 nm) (c=3, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...