Tisk…

Tris(dimethylamino)phosphine 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Tris(dimethylamino)phosphine, Hexamethyl phosphorous triamide, Hexamethyltriaminophosphine, HMPT

162480100 162480500 162481000
ACRO162480100EA 2240 CZK
ACRO162480100 ACRO162480500 ACRO162481000
Tris(dimethylamino)phosphine 97%
Tris(dimethylamino)phosphine
Vzorec: C₆H₁₈N₃P
MW: 163,2 g/mol
Bod varu: 150 °C (760 mmHg)
Bod tání: –44 °C
Hustota: 0,89 g/cm³
Bod vzplanutí: 26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008301
CAS číslo: 1608-26-0
EINECS: 216-534-4
UN: 1993
ADR: 3,III

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
Refractive index 1.4650 to 1.4670 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR