Tisk…

N,N,N-Trimethylanilinium tribromide 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Phenyltrimethylammonium tribromide, Mono(N,N,N-trimethylbenzenaminium) tribromide, Trimethylphenylammonium tribromide

160940250 160941000
ACRO160940250EA 1560 CZK
ACRO160940250 ACRO160941000
N,N,N-Trimethylanilinium tribromide 97%
N,N,N-Trimethylanilinium tribromide
Vzorec: C₉H₁₄Br₃N
MW: 375,93 g/mol
Bod tání: 113…117 °C
MDL Number: MFCD00011789
CAS číslo: 4207-56-1
EINECS: 224-127-8
UN: 3261
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Melting point 113 to 117 °C
Titration ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR