Tisk…

N-ethylmaleimid (NEM) 99+%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: N-Ethylmaleimide, 1-Ethyl-1H-pyrrole-2,5-dione

156100050 156100100 156100500 156102500
ACRO156100050EA 3220 CZK
ACRO156100050 ACRO156100100 ACRO156100500 ACRO156102500
N-ethylmaleimid (NEM) 99+%
N-ethylmaleimid (NEM)
Vzorec: C₆H₇NO₂
MW: 125.13
Bod varu: 210 °C (760 mmHg)
Bod tání: 41…45 °C
Bod vzplanutí: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005509
CAS číslo: 128-53-0
EINECS: 204-892-4
UN: 2928
ADR: 6.1 II
Merck Index: 13,03855

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 41 to 45 °C
Solubility (5% in 100% EtOH) Clear colorless to almost colorless solution
HPLC ≥99.0 %
Residue after ignition ≤0.01 %
Aluminium (Al) ≤1 ppm
Antimony (Sb) ≤1 ppm
Arsenic (As) ≤1 ppm
Boron (B) ≤1 ppm
Calcium (Ca) ≤5 ppm
Chromium (Cr) ≤1 ppm
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Manganese (Mn) ≤1 ppm
Nickel (Ni) ≤1 ppm
Potassium (K) ≤20 ppm
Sodium (Na) ≤20 ppm
Zinc (Zn) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR