Isophthalaldehyde 99%
Dodavatel: Thermo Scientific
Synonyms:
m-Phthalaldehyde, Isophthaldicarboxaldehyde, Isophthaldialdehyde
151740050
151740250
ACRO151740050EA
1500
CZK
ACRO151740050
ACRO151740250
Isophthalaldehyde 99%
Isophthalaldehyde
Vzorec:
C₈H₆O₂ MW: 134,13 g/mol Bod varu: 136 °C (17hPa) Bod tání: 88…90 °C |
MDL Number:
MFCD00003372 CAS číslo: 626-19-7 EINECS: 210-935-8 |
Výsledky specifikačního testu
Appearance | White to light yellow Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 88 to 90 °C |
GC | ≥98.5 % |
Specific optical rotation | -18.0° to -20.0° (20 °C, 589 nm) (c-1, DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...