N-Methylcyclohexylamine 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
N-Cyclohexylmethylamine, Cyclohexyl(methyl)amine
146990050
146992500
146990010
ACRO146990050EA
861
CZK
ACRO146990050
ACRO146992500
ACRO146990010
N-Methylcyclohexylamine 98%
N-Methylcyclohexylamine
Vzorec:
C₇H₁₅N MW: 113,2 g/mol Bod varu: 149 °C (760 mmHg) Bod tání: –8 °C Hustota: 0,86 g/cm³ Bod vzplanutí: 29 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003832 CAS číslo: 100-60-7 EINECS: 202-869-3 UN: 2734 ADR: 8,II |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4535 to 1.4575 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...