Tisk…

5-Methylphenazinium methyl sulphate 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: PMS, 5-Methylphenazin-5-ium methyl sulphate, Phenazine methosulphate, Phenazine methylsulphate

130160010 130160100 130160250
ACRO130160010EA 2770 CZK
ACRO130160010 ACRO130160100 ACRO130160250
5-Methylphenazinium methyl sulphate 98%
5-Methylphenazinium methyl sulphate
Vzorec: C₁₄H₁₄N₂O₄S
MW: 306,34 g/mol
Bod tání: 158…160 °C
Storage Temperature: Freezer
MDL Number: MFCD00011923
CAS číslo: 299-11-6
EINECS: 206-072-1
Merck Index: 12,06185

Objednat

Výsledky specifikačního testu

Appearance Dark yellow to khaki to brown Crystalline powder
Infrared spectrum Conforms
UV spectrophotometry ≥97.5 % (0.01g/l H2O, 1cm cell, lambda max 387.5 nm)
GC ≥96.0 %
UV at 260 nm A: ≤0.02
Titration after Ion exchange ≥99.0 % (on dry substance)
Membrane filtration test (on 0.4 M soln.) Passes test
Suitability (HPLC gradient) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR