Tisk…

Ethyl phenylpropiolate 98%

Dodavatel: Thermo Scientific

Synonyms: Phenylpropiolic acid ethyl ester, Ethyl 3-phenylprop-2-ynoate, Ethyl phenylacetylenecarboxylate

118850050 118850250
ACRO118850050EA 1230 CZK
ACRO118850050 ACRO118850250
Ethyl phenylpropiolate 98%
Ethyl phenylpropiolate
Vzorec: C₁₁H₁₀O₂
MW: 174,2 g/mol
Bod varu: 260 °C (1013 hPa)
Bod tání: 60 °C
Hustota: 1,05 g/cm³
Bod vzplanutí: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009185
CAS číslo: 2216-94-6
EINECS: 218-703-8

Objednat

Výsledky specifikačního testu

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5510 to 1.5530 (20°C, 589 nm)
GC ≥97.5 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR