Tisk…

3-Brompropylamine hydrobromide 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 3-Bromopropan-1-amine hydrobromide, 3-Bromopropylammonium bromide

107380250 107381000
ACRO107380250EA 1780 CZK
ACRO107380250 ACRO107381000
3-Brompropylamine hydrobromide 98%
3-Brompropylamine hydrobromide
Vzorec: BrH₂CCH₂CH₂NH₃Br
MW: 218.92
Bod tání: 171…175 °C
MDL Number: MFCD00012912
CAS číslo: 5003-71-4
EINECS: 225-675-0

Objednat

Výsledky specifikačního testu

Appearance White to light yellow to light beige to light pink Powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
Residue after ignition ≤0.2 %
Calcium (Ca) ≤10 ppm
Iron (Fe) ≤50 ppm
Magnesium (Mg) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm
Nitrilotriacetic acid ≤0.1 % ((HOCOCH4)3N)
Insoluble matter ≤0.005 % (in dilute NH4OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR