Tisk…

trans-4-Hydroxy-L(-)-prolin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH

M-Clarity™ quality level = MQ200

H54409-100G H54409-10G H54409-2.5G H54409-25G
SIALH54409-100GEA 11610 CZK
SIALH54409-100G SIALH54409-10G SIALH54409-2.5G SIALH54409-25G
trans-4-Hydroxy-L(-)-prolin, Sigma-Aldrich®
trans-4-Hydroxy-L(-)-prolin
Vzorec: C₅H₉NO₃
MW: 131,13 g/mol
Bod varu: 296 °C (1013 hPa)
Bod tání: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00064320
CAS číslo: 51-35-4
EINECS: 200-091-9
Merck Index: 13,04866

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR