L-prolin, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
81709-100G
81709-25G
SIAL81709-100GEA
13560
CZK
SIAL81709-100G
SIAL81709-25G
L-prolin, Sigma-Aldrich®
L-prolin
Vzorec:
C₅H₉NO₂ Bod varu: 245 °C (1013 hPa) Bod tání: 220…222 °C Hustota: 1,35 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064318 CAS číslo: 147-85-3 EINECS: 205-702-2 Merck Index: 13,07871 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...