Tisk…

D-(+)-Proline, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: H-D-Pro-OH, (R)-(+)-Proline, D-Proline

M-Clarity™ quality level = MQ200

858919-500MG 858919-5G
SIAL858919-500MGEA 2350 CZK
SIAL858919-500MG SIAL858919-5G
D-(+)-Proline, Sigma-Aldrich®
D-(+)-Proline
Vzorec: C₅H₉NO₂
Bod tání: 223 °C
MDL Number: MFCD00064317
CAS číslo: 344-25-2
EINECS: 206-452-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR