Tisk…

D-(+)-Proline 99+%

Dodavatel: Thermo Fisher Scientific

Synonyms: H-D-Pro-OH, (R)-(+)-Proline, D-Proline

228660010 228660050 228660250
ACRO228660010EA 1520 CZK
ACRO228660010 ACRO228660050 ACRO228660250
D-(+)-Proline 99+%
D-(+)-Proline
Vzorec: C₅H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00064317
CAS číslo: 344-25-2
EINECS: 206-452-7

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C, 3 hrs)
Sulfated ash ≤0.1 %
Solubility (5 % in water) Clear colorless to light yellow
Specific optical rotation +83.0° to +87.0° (20°C, 589 nm) (c=4, water)
Titration with HClO4 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR