(S)-(+)-Mandelic acid ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
L(+)-Mandelic acid, (S)-(+)-2-Hydroxy-2-phenylacetic acid, L(+)-2-Hydroxy-2-phenylacetic acid, (S)-2-Hydroxy-2-phenylacetic acid
A15061.06
A15061.14
A15061.22
A15061.06EA
924
CZK
A15061.06
A15061.14
A15061.22
(S)-(+)-Mandelic acid ≥99%
(S)-(+)-Mandelic acid
Purity: 99+%
CAS No.: 17199-29-0
Molecular Formula: C8H8O3
Formula Weight: 152.15
Melting Point: 131-135°
Beilstein Reference: 2208678
Vzorec:
C₈H₈O₃ Bod varu: 322 °C (1013 hPa) Bod tání: 131…135 °C Hustota: 1,349 g/cm³ (20 °C) Bod vzplanutí: >190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004495 CAS číslo: 17199-29-0 EINECS: 241-240-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...