(R)-(-)-Mandelic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
M-Clarity™ quality level = MQ100
154210-25G
154210-5G
SIAL154210-25GEA
2160
CZK
SIAL154210-25G
SIAL154210-5G
(R)-(-)-Mandelic acid, Sigma-Aldrich®
(R)-(-)-Mandelic acid
Vzorec:
C₈H₈O₃ MW: 152,15 g/mol Bod varu: 322 °C (1013 hPa) Bod tání: 130…132 °C Hustota: 1,341 g/cm³ (20 °C) Bod vzplanutí: >190 °C |
MDL Number:
MFCD00064251 CAS číslo: 611-71-2 EINECS: 210-276-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...