Tisk…

(R)-(-)-Mandelic acid ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid

A14767.14 A14767.22
A14767.14EA 2690 CZK
A14767.14 A14767.22
(R)-(-)-Mandelic acid ≥98%
(R)-(-)-Mandelic acid

Purity: 98% CAS No.: 611-71-2 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 130-134° Beilstein Reference: 2691094

Vzorec: C₈H₈O₃
MW: 152,15 g/mol
Bod varu: 322 °C (1013 hPa)
Bod tání: 130…134 °C
Hustota: 1,341 g/cm³ (20 °C)
Bod vzplanutí: >190 °C
Storage Temperature: Ambient
MDL Number: MFCD00064251
CAS číslo: 611-71-2
EINECS: 210-276-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR