Tisk…

6-Fluoro-4-cyanoquinoline N-oxide 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Fluoro-1-oxidoquinolin-1-ium-4-carbonitrile

PC99144-250MG PC99144-1G
APOSPC99144-250MGEA 6450 CZK
APOSPC99144-250MG APOSPC99144-1G
6-Fluoro-4-cyanoquinoline N-oxide 95%
6-Fluoro-4-cyanoquinoline N-oxide
Vzorec: C₁₀H₅FN₂O
MW: 188.16 g/mol
MDL Number: MFCD33404154
CAS číslo: 152562-07-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR