3-(Cyclopropyl)-2-(hydroxymethyl)thiophene 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR315885-250MG
OR315885-1G
OR315885-5G
APOSOR315885-250MGEA
5390
CZK
APOSOR315885-250MG
APOSOR315885-1G
APOSOR315885-5G
3-(Cyclopropyl)-2-(hydroxymethyl)thiophene 95%
3-(Cyclopropyl)-2-(hydroxymethyl)thiophene
Vzorec:
C₈H₁₀OS Storage Temperature: Refrigerator |
MDL Number:
MFCD32689353 CAS číslo: 2296417-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...