Tisk…

1-(Cyclopropyl)-1H-pyrazole-3-carbaldehyde 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Cyclopropyl-1H-pyrazole-3-carbaldehyde

OR315862-250MG OR315862-1G
APOSOR315862-250MGEA 6850 CZK
APOSOR315862-250MG APOSOR315862-1G
1-(Cyclopropyl)-1H-pyrazole-3-carbaldehyde 95%
1-(Cyclopropyl)-1H-pyrazole-3-carbaldehyde
Vzorec: C₇H₈N₂O
MW: 136.15 g/mol
Storage Temperature: Freezer
MDL Number: MFCD26520323
CAS číslo: 1823349-88-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR