Tisk…

3-Bromothiophene-4-sulphonyl chloride 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR315853-250MG OR315853-1G
APOSOR315853-250MGEA 6220 CZK
APOSOR315853-250MG APOSOR315853-1G
3-Bromothiophene-4-sulphonyl chloride 95%
3-Bromothiophene-4-sulphonyl chloride
Vzorec: C₄H₂BrClO₂S₂
Storage Temperature: Refrigerator
MDL Number: MFCD04115406
CAS číslo: 111283-90-0
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR