Tisk…

3-Bromo-4-(cyclopropyl)phenol 95%

Dodavatel: Apollo Scientific
Varování

OR315845-250MG OR315845-1G OR315845-5G
APOSOR315845-250MGEA 4960 CZK
APOSOR315845-250MG APOSOR315845-1G APOSOR315845-5G
3-Bromo-4-(cyclopropyl)phenol 95%
3-Bromo-4-(cyclopropyl)phenol
Vzorec: C₉H₉BrO
Storage Temperature: Freezer
MDL Number: MFCD32669828
CAS číslo: 1784977-63-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR