Tisk…

2-Bromo-6-(cyclopropyl)aniline 95%

Dodavatel: Apollo Scientific
Varování

OR315840-250MG OR315840-1G OR315840-5G
APOSOR315840-250MGEA 5390 CZK
APOSOR315840-250MG APOSOR315840-1G APOSOR315840-5G
2-Bromo-6-(cyclopropyl)aniline 95%
2-Bromo-6-(cyclopropyl)aniline
Vzorec: C₉H₁₀BrN
Storage Temperature: Freezer
MDL Number: MFCD32669826
CAS číslo: 2640734-79-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR