2-Bromo-6-(cyclopropyl)nitrobenzene 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR315839-250MG
OR315839-1G
OR315839-5G
APOSOR315839-250MGEA
4960
CZK
APOSOR315839-250MG
APOSOR315839-1G
APOSOR315839-5G
2-Bromo-6-(cyclopropyl)nitrobenzene 95%
2-Bromo-6-(cyclopropyl)nitrobenzene
Vzorec:
C₉H₈BrNO₂ Storage Temperature: Freezer |
MDL Number:
MFCD32669825 CAS číslo: 2640734-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...