2-Methyloxetane-2-carboxylic acid 90+%
Dodavatel: Apollo Scientific
|
Varování
|
OR309088-100MG
OR309088-250MG
APOSOR309088-100MGEA
4100
CZK
APOSOR309088-100MG
APOSOR309088-250MG
2-Methyloxetane-2-carboxylic acid 90+%
2-Methyloxetane-2-carboxylic acid
Vzorec:
C₅H₈O₃ Storage Temperature: Freezer |
MDL Number:
MFCD22571611 CAS číslo: 1305207-92-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...