Tisk…

3-Methoxy-2,4,5-trifluoroaniline 95%

Dodavatel: Apollo Scientific
Varování

PC7784-1G PC7784-5G
APOSPC7784-1GEA 17300 CZK
APOSPC7784-1G APOSPC7784-5G
3-Methoxy-2,4,5-trifluoroaniline 95%
3-Methoxy-2,4,5-trifluoroaniline
Vzorec: C₇H₆F₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD03094442
CAS číslo: 114214-45-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR