3-Amino-5-chloro-2-hydroxypyridine 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR019982-5G
OR019982-10G
OR019982-25G
APOSOR019982-5GEA
4960
CZK
APOSOR019982-5G
APOSOR019982-10G
APOSOR019982-25G
3-Amino-5-chloro-2-hydroxypyridine 97%
3-Amino-5-chloro-2-hydroxypyridine
Vzorec:
C₅H₅ClN₂O |
MDL Number:
MFCD07375031 CAS číslo: 98027-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...