Tisk…

2-Bromo-5-fluoro-3-(trifluoromethyl)benzonitrile 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC57803-250MG PC57803-1G PC57803-5G
APOSPC57803-250MGEA 3540 CZK
APOSPC57803-250MG APOSPC57803-1G APOSPC57803-5G
2-Bromo-5-fluoro-3-(trifluoromethyl)benzonitrile 95%
2-Bromo-5-fluoro-3-(trifluoromethyl)benzonitrile
Vzorec: C₈H₂BrF₄N
MDL Number: MFCD32903690
CAS číslo: 2090965-03-8
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR