2-Bromo-5-(difluoromethyl)benzyl bromide 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC57773-250MG
PC57773-1G
PC57773-5G
APOSPC57773-250MGEA
5460
CZK
APOSPC57773-250MG
APOSPC57773-1G
APOSPC57773-5G
2-Bromo-5-(difluoromethyl)benzyl bromide 95%
2-Bromo-5-(difluoromethyl)benzyl bromide
Vzorec:
C₈H₆Br₂F₂ |
MDL Number:
MFCD18391069 CAS číslo: 1261441-93-3 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...