2,4-Difluoro-3-(trifluoromethoxy)phenol
Dodavatel: Apollo Scientific
Varování
|
PC57486-1G
PC57486-5G
APOSPC57486-1GEA
16340
CZK
APOSPC57486-1G
APOSPC57486-5G
2,4-Difluoro-3-(trifluoromethoxy)phenol
2,4-Difluoro-3-(trifluoromethoxy)phenol
Vzorec:
C₇H₃F₅O₂ |
MDL Number:
MFCD34595684 CAS číslo: 2383762-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...