Tisk…

Methyl 2-(difluoromethoxy)-5-fluorobenzoate

Dodavatel: Apollo Scientific
Varování

PC57483-1G PC57483-5G
APOSPC57483-1GEA 17360 CZK
APOSPC57483-1G APOSPC57483-5G
Methyl 2-(difluoromethoxy)-5-fluorobenzoate
Methyl 2-(difluoromethoxy)-5-fluorobenzoate
Vzorec: C₉H₇F₃O₃
MDL Number: MFCD18399132
CAS číslo: 1261879-66-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR