2-Fluoro-4-iodo-3-methoxypyridine
Dodavatel: Apollo Scientific
|
Varování
|
PC57451-500MG
PC57451-1G
APOSPC57451-500MGEA
8270
CZK
APOSPC57451-500MG
APOSPC57451-1G
2-Fluoro-4-iodo-3-methoxypyridine
2-Fluoro-4-iodo-3-methoxypyridine
Vzorec:
C₆H₅FINO MW: 253.013 g/mol |
MDL Number:
MFCD16611136 CAS číslo: 1227580-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...