Tisk…

2'-Bromo-3',6',2,2,2-pentafluoroacetophenone 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(2-Bromo-3,6-difluorophenyl)-2,2,2-trifluoroethanone

PC57434-250MG PC57434-1G
APOSPC57434-250MGEA 6220 CZK
APOSPC57434-250MG APOSPC57434-1G
2'-Bromo-3',6',2,2,2-pentafluoroacetophenone 95%
2'-Bromo-3',6',2,2,2-pentafluoroacetophenone
Vzorec: C₈H₂BrF₅O
MW: 288.9968 g/mol
MDL Number: MFCD12922615
CAS číslo: 1208076-54-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR