Tisk…

4-Bromo-2-hydroxy-3-(trifluoromethyl)benzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC57420-250MG PC57420-1G PC57420-5G
APOSPC57420-250MGEA 4960 CZK
APOSPC57420-250MG APOSPC57420-1G APOSPC57420-5G
4-Bromo-2-hydroxy-3-(trifluoromethyl)benzoic acid 95%
4-Bromo-2-hydroxy-3-(trifluoromethyl)benzoic acid
Vzorec: C₈H₄BrF₃O₃
MW: 285.01 g/mol
CAS číslo: 2385518-14-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR