1-Bromo-3,5-dichloro-4-fluoro-2-iodobenzene 95%
Dodavatel: Apollo Scientific
Nebezpečí
|
PC57411-250MG
PC57411-1G
PC57411-5G
APOSPC57411-250MGEA
5390
CZK
APOSPC57411-250MG
APOSPC57411-1G
APOSPC57411-5G
1-Bromo-3,5-dichloro-4-fluoro-2-iodobenzene 95%
1-Bromo-3,5-dichloro-4-fluoro-2-iodobenzene
Vzorec:
C₆HBrCl₂FI MW: 369.79 g/mol |
MDL Number:
MFCD34166239 CAS číslo: 2676196-06-6 UN: 2811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...