Tisk…

5-Chloro-4-fluoro-2-(trifluoromethyl)benzonitrile 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC56987-250MG PC56987-1G PC56987-5G
APOSPC56987-250MGEA 4960 CZK
APOSPC56987-250MG APOSPC56987-1G APOSPC56987-5G
5-Chloro-4-fluoro-2-(trifluoromethyl)benzonitrile 95%
5-Chloro-4-fluoro-2-(trifluoromethyl)benzonitrile
Vzorec: C₈H₂ClF₄N
MDL Number: MFCD28752675
CAS číslo: 1807257-71-1
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR