3-(1,1-Difluoroethyl)benzaldehyde 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC52529-250MG
PC52529-1G
PC52529-5G
APOSPC52529-250MGEA
5390
CZK
APOSPC52529-250MG
APOSPC52529-1G
APOSPC52529-5G
3-(1,1-Difluoroethyl)benzaldehyde 95%
3-(1,1-Difluoroethyl)benzaldehyde
Vzorec:
C₉H₈F₂O |
MDL Number:
MFCD20527725 CAS číslo: 1032527-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...