6-(Trifluoromethoxy)-2-(trifluoromethyl)-1H-indole 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC52229-250MG
PC52229-1G
APOSPC52229-250MGEA
6220
CZK
APOSPC52229-250MG
APOSPC52229-1G
6-(Trifluoromethoxy)-2-(trifluoromethyl)-1H-indole 95%
6-(Trifluoromethoxy)-2-(trifluoromethyl)-1H-indole
Vzorec:
C₁₀H₅F₆NO MW: 269,14 g/mol |
MDL Number:
MFCD20226878 CAS číslo: 1067188-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...