3-Bromoimidazo[1,2-a]pyridine-8-carbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Bromo-2,3-dihydroimidazo[1,2-a]pyridine-8-carbonitrile
OR70193-250MG
OR70193-1G
APOSOR70193-250MGEA
3940
CZK
APOSOR70193-250MG
APOSOR70193-1G
3-Bromoimidazo[1,2-a]pyridine-8-carbonitrile
3-Bromoimidazo[1,2-a]pyridine-8-carbonitrile
Vzorec:
C₈H₆BrN₃ |
MDL Number:
MFCD28154606 CAS číslo: 1383718-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...