3-Bromo-7-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR47911-250MG
OR47911-1G
APOSOR47911-250MGEA
3940
CZK
APOSOR47911-250MG
APOSOR47911-1G
3-Bromo-7-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
3-Bromo-7-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Vzorec:
C₈H₄BrClN₂O₂ |
MDL Number:
MFCD32857213 CAS číslo: 2594436-02-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...