2-Bromo-5-chloro-4-fluorobenzyl alcohol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2-Bromo-5-chloro-4-fluorophenyl)methanol
PC50331-1G
PC50331-5G
APOSPC50331-1GEA
10550
CZK
APOSPC50331-1G
APOSPC50331-5G
2-Bromo-5-chloro-4-fluorobenzyl alcohol
2-Bromo-5-chloro-4-fluorobenzyl alcohol
Vzorec:
C₇H₅BrClFO MW: 239,47 g/mol |
MDL Number:
MFCD30297360 CAS číslo: 1067882-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...