Tisk…

2-Bromo-5-chloro-4-fluorobenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

PC50306-1G PC50306-5G
APOSPC50306-1GEA 17360 CZK
APOSPC50306-1G APOSPC50306-5G
2-Bromo-5-chloro-4-fluorobenzyl bromide
2-Bromo-5-chloro-4-fluorobenzyl bromide
Vzorec: C₇H₄Br₂ClF
MW: 302,37 g/mol
MDL Number: MFCD30296109
CAS číslo: 1067882-52-3
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR