Tisk…

Ethyl 4-amino-6-chloro-5-fluoronicotinate 99%

Dodavatel: Apollo Scientific
Varování

PC99415-1G PC99415-5G PC99415-10G
APOSPC99415-1GEA 4200 CZK
APOSPC99415-1G APOSPC99415-5G APOSPC99415-10G
Ethyl 4-amino-6-chloro-5-fluoronicotinate 99%
Ethyl 4-amino-6-chloro-5-fluoronicotinate
Vzorec: C₈H₈ClFN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD34471315
CAS číslo: 2454397-74-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR