meso-2,3-Dimercaptosuccinic acid, Sigma-Aldrich®
Dodavatel: Merck
D7881-1G
D7881-25G
D7881-5G
D7881-50G
SIALD7881-1GEA
1230
CZK
SIALD7881-1G
SIALD7881-25G
SIALD7881-5G
SIALD7881-50G
meso-2,3-Dimercaptosuccinic acid, Sigma-Aldrich®
meso-2,3-Dimercaptosuccinic acid
Vzorec:
C₄H₆O₄S₂ Bod tání: 210…211 °C Storage Temperature: Freezer |
MDL Number:
MFCD00064799 CAS číslo: 304-55-2 EINECS: 206-155-2 Merck Index: 12,09034 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...