Tisk…

Ethyl 3-methyl-6-nitroimidazo[1,2-a]pyridine-2-carboxylate

Dodavatel: Apollo Scientific
Varování

OR111572-250MG OR111572-1G
APOSOR111572-250MGEA 4960 CZK
APOSOR111572-250MG APOSOR111572-1G
Ethyl 3-methyl-6-nitroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-methyl-6-nitroimidazo[1,2-a]pyridine-2-carboxylate
Vzorec: C₁₁H₁₁N₃O₄
MDL Number: MFCD31629940
CAS číslo: 1612861-50-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR