2,6-Difluoro-3-(trifluoromethoxy)benzyl bromide 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC502457-250MG
PC502457-1G
PC502457-5G
APOSPC502457-250MGEA
4960
CZK
APOSPC502457-250MG
APOSPC502457-1G
APOSPC502457-5G
2,6-Difluoro-3-(trifluoromethoxy)benzyl bromide 95%
2,6-Difluoro-3-(trifluoromethoxy)benzyl bromide
Vzorec:
C₈H₄BrF₅O MW: 291.01 g/mol |
MDL Number:
MFCD31561157 CAS číslo: 2244081-55-6 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...